Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TCR beta Monoclonal Antibody (H57-597), Brilliant Ultra Violet™ 661, eBioscience™, Invitrogen™
Armenian Hamster Monoclonal Antibody
$215.00 - $437.00
Specifications
Antigen | TCR beta |
---|---|
Clone | H57-597 |
Concentration | 0.2 mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
37-659-6182
![]() |
Invitrogen™
376596182 |
100 μg |
Each for $437.00
|
|
|||||
37-659-6180
![]() |
Invitrogen™
376596180 |
25 μg |
Each for $215.00
|
|
|||||
Description
TCR beta Monoclonal antibody specifically detects TCR beta in Mouse samples. It is validated for Flow CytometrySpecifications
TCR beta | |
0.2 mg/mL | |
Monoclonal | |
Armenian Hamster | |
Mouse | |
bTCR; FLJ22602; MGC117436; MGC22624; MGC23964; MGC71411; T cell receptor beta chain; t-cell antigen receptor; T-cell receptor; TCR beta; Tcra; Tcrb; TCRbeta; TCRD; Tib; TRA | |
IgG | |
Affinity chromatography | |
Tcrb |
H57-597 | |
Flow Cytometry | |
Brilliant Ultraviolet 661 | |
RUO | |
PBS with BSA and 0.09% sodium azide; pH 7.2 | |
21577 | |
Primary | |
4°C, store in dark, DO NOT FREEZE! |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title