Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TCR Cb1 Mouse anti-Human, Brilliant Violet 711, Clone: JOVI.1, BD OptiBuild™
Mouse Monoclonal Antibody
Supplier: BD Biosciences 747977
Description
TCR Cb1 Monoclonal antibody specifically detects TCR Cb1 in Human samples. It is validated for Flow Cytometry.
Specifications
TCR Cb1 | |
Monoclonal | |
0.2 mg/mL | |
Mouse | |
RUO | |
TCR Cb1 | |
Refrigerate (2 to 8°C) |
Flow Cytometry | |
JOVI.1 | |
Brilliant Violet 711 | |
50 μg | |
Primary | |
Human | |
IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction