Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TCR β Chain Hamster anti-Mouse, Brilliant Ultraviolet 737, Clone: H57-597, BD Horizon™
Hamster Monoclonal Antibody
Supplier: BD Biosciences 612821
Description
TCR β Chain Monoclonal antibody specifically detects TCR β Chain in samples. It is validated for Flow cytometry.
Specifications
TCR β Chain | |
Monoclonal | |
0.2mg/mL | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
TCR affinity-purified from mouse T-cell hybridoma DO-11.10 | |
50 μg | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Flow Cytometry | |
H57-597 | |
Brilliant Ultraviolet 737 | |
Hamster | |
Affinity Purified | |
RUO | |
Mouse | |
IgG2 |
Safety and Handling
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction