Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TCR DO11.10 Monoclonal Antibody (JKJ1-26), Abbomax

Catalog No. 502204437
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-220-4437 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-220-4437 Supplier Abbomax Inc Supplier No. 606330
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Positive control: Tonsil Cellular location: Membrane.

TCR DO11.10 Monoclonal antibody specifically detects TCR DO11.10 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry (Paraffin)

Specifications

Antigen TCR DO11.10
Applications Flow Cytometry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone JKJ1-26
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with proprietary stabilizer and 0.01% sodium azide
Gene Alias D011.10; T cell receptor alpha chain; Tcra; Tcralpha
Host Species Mouse
Immunogen DO11.10 TCR hybridoma cells
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.