Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TCR DO11.10 Monoclonal Antibody (KJ1-26), Biotin
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA517661
Description
MA5-17661 targets D011.10 TCR in FACS applications and shows reactivity with Mouse samples. The MA5-17661 immunogen is t cell hybridoma, D011.10.
TCR DO11 (DO11.10) is a receptor found on lymphocytes of DO11.10 transgenic mice and T-cell receptors of BALB/c-derived DO11.10 and DO11.10.24 T cell hybridomas. The DO11.10 TCR recognizes a C-terminal ovalbumin peptide (OVA 323-339) in the context of I-Ad major histocompatibility molecules. The use of antibodies against DO11.10 have proven to be instrumental in describing many of the intricate details of T cell immunity, including T cell activation, development, and function.
Specifications
TCR DO11.10 | |
Monoclonal | |
0.1 mg/mL | |
PBS with 4-5mg/mL BSA and 0.02% sodium azide | |
P01849 | |
Tcra | |
T cell hybridoma, D011.10. | |
100 μg | |
Primary | |
Mouse | |
Antibody | |
IgG2a |
Flow Cytometry | |
KJ1-26 | |
Biotin | |
Tcra | |
D011.10; T cell receptor alpha chain; Tcra; Tcralpha | |
Mouse | |
Protein G | |
RUO | |
21473 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction