Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TCR DO11.10 Monoclonal Antibody (KJ1-26 (KJ126, KJI-26, KJI26)), Biotin, eBioscience™, Invitrogen™
GREENER_CHOICE

Catalog No. 5011932
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-119-32 100 μg
50-119-31 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-119-32 Supplier Invitrogen™ Supplier No. 13580882
Add to Cart
Edge
Add to Cart

Mouse Monoclonal Antibody

Description: The KJ1-26 anti-idiotypic monoclonal antibody binds the T-cell receptor (TCR) expressed on DO11.10 and DO11.10.24 murine T cell hybridomas, but not on any other T cell hybridomas tested.The TCR expressed on DO11.10 recognizes an ovalbumin peptide in the context of I-A^{d^} major histocompatibility molecules.The use of DO11.10 and KJ1-26 have proven to be instrumental in describing many of the intricate details of T cell immunity. Applications Reported: This KJ1-26 (KJ126, KJI-26, KJI26) antibody has been reported for use in flow cytometric analysis. Applications Tested: This KJ1-26 (KJ126, KJI-26, KJI26) antibody has been tested by flow cytometric analysis of the DO11.10 T cell hybridoma. This can be used at less than or equal to 0.25 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Filtration: 0.2 μm post-manufacturing filtered.

TCR DO11 (DO11.10) is a receptor found on lymphocytes of DO11.10 transgenic mice and T-cell receptors of BALB/c-derived DO11.10 and DO11.10.24 T cell hybridomas. The DO11.10 TCR recognizes a C-terminal ovalbumin peptide (OVA 323-339) in the context of I-Ad major histocompatibility molecules. The use of antibodies against DO11.10 have proven to be instrumental in describing many of the intricate details of T cell immunity, including T cell activation, development, and function.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TCR DO11.10
Applications Flow Cytometry
Classification Monoclonal
Clone KJ1-26 (KJ126, KJI-26, KJI26), KJI-26, KJI26)
Concentration 0.5 mg/mL
Conjugate Biotin
Formulation PBS with 0.09% sodium azide; pH 7.2
Gene Tcra
Gene Accession No. P01849
Gene Alias D011.10; T cell receptor alpha chain; Tcra; Tcralpha
Gene Symbols Tcra
Host Species Mouse
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 21473
Target Species Mouse
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.