Learn More
Invitrogen™ TCR DO11.10 Monoclonal Antibody (KJ1-26), PE

Description
MA5-17663 targets D011.10 TCR in FACS applications and shows reactivity with Mouse samples. The MA5-17663 immunogen is t cell hybridoma, D011.10.
Specifications
Specifications
| Antigen | TCR DO11.10 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | KJ1-26 |
| Concentration | 0.1 mg/mL |
| Conjugate | PE |
| Formulation | PBS with 4-5mg/mL BSA and 0.02% sodium azide |
| Gene | Tcra |
| Gene Accession No. | P01849 |
| Gene Alias | D011.10; T cell receptor alpha chain; Tcra; Tcralpha |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.