Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TCR DO11.10 Monoclonal Antibody (KJ1-26), PE-Cyanine5
GREENER_CHOICE

Catalog No. PIMA517664
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA517664 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA517664 Supplier Invitrogen™ Supplier No. MA517664
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MA5-17664 targets D011.10 TCR in FACS applications and shows reactivity with Mouse samples. The MA5-17664 immunogen is t cell hybridoma, D011.10.

TCR DO11 (DO11.10) is a receptor found on lymphocytes of DO11.10 transgenic mice and T-cell receptors of BALB/c-derived DO11.10 and DO11.10.24 T cell hybridomas. The DO11.10 TCR recognizes a C-terminal ovalbumin peptide (OVA 323-339) in the context of I-Ad major histocompatibility molecules. The use of antibodies against DO11.10 have proven to be instrumental in describing many of the intricate details of T cell immunity, including T cell activation, development, and function.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TCR DO11.10
Applications Flow Cytometry
Classification Monoclonal
Clone KJ1-26
Concentration 0.1 mg/mL
Conjugate PE-Cyanine5
Formulation PBS with 4-5mg/mL BSA and 0.02% sodium azide
Gene Tcra
Gene Accession No. P01849
Gene Alias D011.10; T cell receptor alpha chain; Tcra; Tcralpha
Gene Symbols Tcra
Host Species Mouse
Immunogen T cell hybridoma, D011.10.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 21473
Target Species Mouse
Content And Storage 4°C, do not freeze
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.