Learn More
Invitrogen™ TCR DO11.10 Monoclonal Antibody (KJ126.1)

Description
This antibody is specific for the DO-11.10 T cell receptor idiotype. The DO-11.10 TCR is specific for chicken ovalbumin (aa 323-339) in combination with I-Ad MHC class II complex. KJ1-26 has been reported to block the antigen response of the DO-11.10 T cell hybridomas in vitro.
Specifications
Specifications
| Antigen | TCR DO11.10 |
| Applications | ELISA, Flow Cytometry |
| Classification | Monoclonal |
| Clone | KJ126.1 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.1% sodium azide |
| Gene | Tcra |
| Gene Accession No. | P01849 |
| Gene Alias | D011.10; T cell receptor alpha chain; Tcra; Tcralpha |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.