Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TCR gamma/delta Antibody (B1), mFluor Violet 500 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP262225MFV500
Description
TCR gamma/delta Monoclonal antibody specifically detects TCR gamma/delta in Human, Primate samples. It is validated for Flow Cytometry, Flow (Cell Surface)Specifications
TCR gamma/delta | |
Monoclonal | |
mFluor Violet 500 SE | |
50mM Sodium Borate | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human, Primate | |
Purified |
Flow Cytometry | |
B1 | |
Flow Cytometry, Flow (Cell Surface) | |
T cell receptor gamma locus, T-cell antigen receptor, gamma polypeptide, T-cell rearranging gene, gamma, T-cell receptor, gamma cluster, TCRG, TRG | |
TCR gamma/delta, the subtype of T cell receptor expressed mainly in epithelial tissues and at the sites of infection. | |
0.1 mL | |
Adaptive Immunity, Immunology, Stem Cells | |
6965 | |
Store at 4°C in the dark. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction