Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TCR gamma/delta Antibody (GL-3) [DyLight 755], Novus Biologicals™

Hamster Monoclonal Antibody
Supplier: Novus Biologicals FAB7297Z
Description
TCR gamma/delta Monoclonal antibody specifically detects TCR gamma/delta in Mouse samples. It is validated for Flow Cytometry, CyTOF-reportedSpecifications
TCR gamma/delta | |
Monoclonal | |
DyLight 755 | |
50mM Sodium Borate | |
Hamster | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Mouse | |
Purified |
Flow Cytometry, CyTOF | |
GL-3 | |
Flow Cytometry, CyTOF-reported | |
T cell receptor gamma locus, T-cell antigen receptor, gamma polypeptide, T-cell rearranging gene, gamma, T-cell receptor, gamma cluster, TCRG, TRG | |
Mouse intraepithelial lymphocytes | |
0.1 mL | |
Adaptive Immunity, Immunology, Stem Cells | |
6965 | |
Store at 4°C in the dark. | |
IgG2 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction