Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TCR gamma/delta Monoclonal Antibody (TCR-1)
GREENER_CHOICE

Catalog No. PIMA528511
Change view
Click to view available options
Quantity:
250 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA528511 250 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA528511 Supplier Thermo Scientific Supplier No. MA528511
Add to Cart
Add to Cart

TCR gamma/delta Monoclonal Antibody (TCR-1)

TCR gamma/delta Monoclonal Antibody for IHC (F), Flow, IP

TRUSTED_SUSTAINABILITY

Specifications

Antigen TCR gamma/delta
Applications Flow Cytometry, Immunohistochemistry (Frozen), Immunoprecipitation
Classification Monoclonal
Clone TCR-1
Concentration 500 μg/mL
Conjugate Unconjugated
Formulation BBS with <0.1% sodium azide
Gene Alias RGD1311300; similar to T cell receptor V delta 6; T cell receptor gamma locus; T3/TCR complex; TCR delta gamma; TCR gamma/delta; TCR-CD3 complex; Tcrg; Trg; Trg@
Host Species Mouse
Purification Method Ion-exchange chromatography
Quantity 250 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Chicken
Content And Storage 4°C, do not freeze
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.