Learn More
Invitrogen™ TCR gamma/delta Monoclonal Antibody (UC7-13D5), Alexa Fluor™ 488

Description
TCR gamma/delta Monoclonal Antibody for Flow
Specifications
Specifications
| Antigen | TCR gamma/delta |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | UC7-13D5 |
| Concentration | 0.5 mg/mL |
| Conjugate | Alexa Fluor 488 |
| Formulation | PBS with <0.1% sodium azide; pH 7.4 |
| Gene | Tcrg |
| Gene Accession No. | 0 |
| Gene Alias | RGD1311300; similar to T cell receptor V delta 6; T cell receptor gamma locus; T3/TCR complex; TCR delta gamma; TCR gamma/delta; TCR-CD3 complex; Tcrg; Trg; Trg@ |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.