Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TCR gamma/delta Antibody (B1), PE/Atto594, Novus Biologicals™
SDP

Catalog No. N262225P594 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N262225P594 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N262225P594 Supplier Novus Biologicals Supplier No. NBP262225PEATT594
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

TCR gamma/delta Monoclonal specifically detects TCR gamma/delta in Human, Primate samples. It is validated for Flow Cytometry, Flow (Cell Surface).

Specifications

Antigen TCR gamma/delta
Applications Flow Cytometry
Classification Monoclonal
Clone B1
Conjugate PE/ATTO 594
Dilution Flow Cytometry, Flow (Cell Surface)
Formulation PBS with 0.05% Sodium Azide
Gene Alias T cell receptor gamma locus, T-cell antigen receptor, gamma polypeptide, T-cell rearranging gene, gamma, T-cell receptor, gamma cluster, TCRG, TRG
Gene Symbols TRG
Host Species Mouse
Immunogen TCR gamma/delta, the subtype of T cell receptor expressed mainly in epithelial tissues and at the sites of infection.
Purification Method Protein A purified
Quantity 0.1 mL
Research Discipline Adaptive Immunity, Immunology, Stem Cells
Primary or Secondary Primary
Gene ID (Entrez) 6965.0
Test Specificity The mouse monoclonal antibody B1 (also known as B1.1) recognizes TCR gamma/delta, the subtype of T cell receptor expressed mainly in epithelial tissues and at the sites of infection.
Target Species Human, Primate
Content And Storage Store at 4°C in the dark. Do not freeze.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.