Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TCRαβ Mouse anti-Human, BV421, Clone: IP26 (also known as IP26A), BD OptiBuild™

Catalog No. BDB744778 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB744778 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB744778 Supplier BD Biosciences Supplier No. 744778
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

TCRαβ Monoclonal antibody specifically detects TCRαβ in Human samples. It is validated for Flow Cytometry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TCRαβ
Applications Flow Cytometry
Classification Monoclonal
Clone IP26 (also known as IP26A)
Concentration 0.2 mg/mL
Conjugate Brilliant Violet 421
Host Species Mouse
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Refrigerate (2 to 8°C)
Form Purified
Isotype IgG1 κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.