Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TCR Vγ1.1 Armenian Hamster anti-Mouse, Unconjugated, Clone: , MilliporeSigma™

Catalog No. MABF93025UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABF930-25UG 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABF930-25UG Supplier MilliporeSigma Supplier No. MABF93025UG
Add to Cart
Add to Cart

Armenian Hamster Monoclonal Antibody

Anti-TCR V 1.1, clone 2.11, Cat. No. MABF930, is an armenian hamster monoclonal antibody that detects T-cell receptor gamma chain C region C10.5. and has been tested for use Flow Cytometry, and Immunoprecipitation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TCR Vγ1.1
Applications Flow Cytometry, Immunoprecipitation
Classification Monoclonal
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Formulation Purified armenian hamster monoclonal antibody IgG in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.
Gene Alias T-cell receptor gamma chain C region C10.5
Host Species Armenian Hamster
Immunogen Irradiated TCR gamma/delta expressing T3 .13.1 hybridoma cells.
Molecular Weight of Antigen 19.10 kDa calculated.
Purification Method Protein G purified
Quantity 25 μg
Regulatory Status RUO
Research Discipline Inflammation & Immunology
Primary or Secondary Primary
Test Specificity Clone 2.11 detects T-cell receptor gamma chain C region C10.5 in lymphocytes.
Target Species Mouse
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Product Type Antibodies
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.