Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TCR Vβ23 Mouse anti-Human, BV421, Clone: AHUT7 (also known as AF23; AF-23; αHUT7; HUT-78; HUT78#7), BD Horizon™

Catalog No. BDB569639 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB569639 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB569639 Supplier BD Biosciences Supplier No. 569639
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

TCR Vβ23 Monoclonal antibody specifically detects TCR Vβ23 in Human samples. It is validated for Flow cytometry
TRUSTED_SUSTAINABILITY

Specifications

Antigen TCR Vβ23
Applications Flow Cytometry
Classification Monoclonal
Clone AHUT7 (also known as AF23; AF-23; αHUT7; HUT-78; HUT78#7)
Conjugate Brilliant Violet 421
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Gene Alias TCR V beta 23; TCRBV23S1; TRBV13; TRBV23-1
Host Species Mouse
Immunogen HUT78 T Cell Leukemia Line
Purification Method Affinity Chromatography
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions and unconjugated antibody and free dye were removed.
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.