Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TCR Vγ4 Mouse anti-Mouse, BV421, Clone: 49.2 (also known as 49.2-9), BD Horizon™
Mouse Monoclonal Antibody
$130.00 - $326.00
Specifications
Antigen | TCR Vγ4 |
---|---|
Clone | 49.2 (also known as 49.2-9) |
Concentration | 0.2 mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Description
TCR Vγ4 Monoclonal antibody specifically detects TCR Vγ4 in Mouse samples. It is validated for Flow cytometrySpecifications
TCR Vγ4 | |
0.2 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
21638 | |
IgG1 κ | |
Affinity Chromatography |
49.2 (also known as 49.2-9) | |
Flow Cytometry | |
Brilliant Violet 421 | |
Mouse | |
Mouse | |
T Cell Receptor Vγ4; TCR Vg4; TCR Vgamma4; TCR Vγ2 (Garman Nomenclature); Tcrg-V4; Trgv4 | |
Mouse TCR γδ Hybridoma Cells | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions and unconjugated antibody and free dye were removed. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title