Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TCR Vγ4 Mouse anti-Mouse, PE, Clone: 49.2 (also known as 49.2-9), BD Phrmingen™
Mouse Monoclonal Antibody
Supplier: BD Biosciences 569159
Description
TCR Vγ4 Monoclonal antibody specifically detects TCR Vγ4 in Mouse samples. It is validated for Flow cytometry
Specifications
TCR Vγ4 | |
Monoclonal | |
0.2 mg/mL | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Mouse | |
Affinity Chromatography | |
RUO | |
21638 | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions and unconjugated antibody and free dye were removed. | |
IgG1 κ |
Flow Cytometry | |
49.2 (also known as 49.2-9) | |
PE | |
T Cell Receptor Vγ4; TCR Vg4; TCR Vgamma4; TCR Vγ2 (Garman Nomenclature); Tcrg-V4; Trgv4 | |
Mouse TCR γδ Hybridoma Cells | |
25 μg | |
Primary | |
Mouse | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction