Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TCR Vβ5.1 Mouse anti-Human, BV750, Clone: LC4, BD OptiBuild™

Catalog No. BDB751800 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
1 x 50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB751800 1 x 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB751800 Supplier BD Biosciences Supplier No. 751800
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

TCR Vβ5.1 Monoclonal antibody specifically detects TCR Vβ5.1 in Human samples. It is validated for Flow Cytometry.

TRUSTED_SUSTAINABILITY

Specifications

Antigen TCR Vβ5.1
Applications Flow Cytometry
Classification Monoclonal
Clone LC4
Concentration 0.2 mg/mL
Conjugate Brilliant Violet 750
Description Hu TCR VBta5.1 BV750 LC4 50 μg
Host Species Mouse
Immunogen Human T acute lymphoblastic leukemia Cell Line
Quantity 1 x 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity TCR Vβ5.1
Target Species Human
Content And Storage Refrigerate (2 to 8°C)
Isotype IgG1 κ
Reactivity Human
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.