Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TCR Vβ5.1 Mouse anti-Human , PE, Clone: LC4, BD Biosciences™
Mouse Monoclonal Antibody
$154.00 - $246.00
Specifications
Antigen | TCR Vβ5.1 |
---|---|
Clone | LC4 |
Applications | Flow Cytometry |
Classification | Monoclonal |
Conjugate | PE |
Description
TCR Vβ5.1 Monoclonal antibody specifically detects TCR Vβ5.1 in Human samples. It is validated for Flow CytometrySpecifications
TCR Vβ5.1 | |
Flow Cytometry | |
PE | |
RUO | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
Human T acute lymphoblastic leukemia Cell Line | |
Primary |
LC4 | |
Monoclonal | |
Mouse | |
Human | |
TCR V beta 5.1; TCR Vb5.1; TRBV5-1; TRBV51 | |
IgG1 κ | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with R-PE under optimum conditions, and unconjugated antibody and free PE were removed. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title