Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TCR Vβ5.2/5.3 Mouse anti-Human, BV 750, Clone: MH3-2, BD OptiBuild™

Catalog No. BDB752679 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB752679 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. BDB752679 Supplier BD Biosciences Supplier No. 752679
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

TCR Vβ5.2/5.3 Monoclonal antibody specifically detects TCR Vβ5.2/5.3 in Human samples. It is validated for Flow Cytometry
TRUSTED_SUSTAINABILITY

Specifications

Antigen TCR Vβ5.2/5.3
Applications Flow Cytometry
Classification Monoclonal
Clone MH3-2
Concentration 0.2 mg/mL
Conjugate Brilliant Violet 750
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Gene Alias TCR Vβ5.2, TRBrilliant Violet 5-6, TCRBrilliant Violet 5S2; TCR Vβ5.3, TRBrilliant Violet 5-5, TCRBrilliant Violet 5S3
Host Species Mouse
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions that minimize unconjugated dye and antibody.
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.