Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TCR Vβ5 Related Mouse anti-Human, Alexa Fluor™ 488, Clone: MEM-262, BD Phrmingen™
Mouse Monoclonal Antibody
$273.50
Specifications
Antigen | TCR Vβ5 Related |
---|---|
Clone | MEM-262 |
Applications | Flow Cytometry |
Classification | Monoclonal |
Conjugate | Alexa Fluor 488 |
Description
TCR Vβ5 Related Monoclonal antibody specifically detects TCR Vβ5 Related in Human samples. It is validated for Flow cytometrySpecifications
TCR Vβ5 Related | |
Flow Cytometry | |
Alexa Fluor 488 | |
Mouse | |
Human | |
MEM-262 Antigen; TCR Vβ5.3 Related; TCR Vb5; TCR Vbeta5; TCRV beta 5 | |
TCR Vβ5.3+ Human HPB-ALL Cell Line | |
Primary | |
The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions and unreacted dye was removed. Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
MEM-262 | |
Monoclonal | |
Liquid | |
RUO | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
6957 | |
IgG2a κ | |
Affinity Chromatography |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title