Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TCR Vα7.2 Mouse anti-Human, PE, Clone: OF-5A12, BD Pharmingen™

Catalog No. BDB566739 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB566739 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB566739 Supplier BD Biosciences Supplier No. 566739
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

TCR Vα7.2 Monoclonal antibody specifically detects TCR Vα7.2 in Human samples. It is validated for Flow Cytometry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TCR Vα7.2
Applications Flow Cytometry
Classification Monoclonal
Clone OF-5A12
Conjugate PE
Host Species Mouse
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity TCR Vα7.2
Target Species Human
Content And Storage Refrigerate (2 to 8°C)
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.