Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TCR Vγ7 Mouse anti-Mouse, BV421, Clone: F2.67 (also known as F4.67), BD Horizon™

Catalog No. BDB569447 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB569447 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB569447 Supplier BD Biosciences Supplier No. 569447
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

TCR Vγ7 Monoclonal antibody specifically detects TCR Vγ7 in Mouse samples. It is validated for Flow cytometry
TRUSTED_SUSTAINABILITY

Specifications

Antigen TCR Vγ7
Applications Flow Cytometry
Classification Monoclonal
Clone F2.67 (also known as F4.67)
Concentration 0.2 mg/mL
Conjugate Brilliant Violet 421
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Gene Alias T Cell Receptor Vγ7; TCR Vg7; TCR Vgamma7; Tcrg-V7; Trgv7
Host Species Mouse
Immunogen Mouse γδ T Cell Hybridoma Cells
Purification Method Affinity Chromatography
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 21641
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions and unconjugated antibody and free dye were removed.
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.