Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TCR Vγ7 Mouse anti-Mouse, PE, Clone: F2.67 (also known as F4.67), BD Phrmingen™
Mouse Monoclonal Antibody
Supplier: BD Biosciences 569155
Description
TCR Vγ7 Monoclonal antibody specifically detects TCR Vγ7 in Mouse samples. It is validated for Flow cytometry
Specifications
| TCR Vγ7 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 21641 | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions and unconjugated antibody and free dye were removed. | |
| IgG2a κ |
| Flow Cytometry | |
| F2.67 (also known as F4.67) | |
| PE | |
| T Cell Receptor Vγ7; TCR Vg7; TCR Vgamma7; Tcrg-V7; Trgv7 | |
| Mouse γδ T Cell Hybridoma Cells | |
| 25 μg | |
| Primary | |
| Mouse | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction