Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TCR V alpha 11 Chimeric Recombinant Rabbit Monoclonal Antibody (34)
Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA547990
Description
Specificity: This antibody binds to the T-cell receptor alpha chains.
The ability of T cell receptors (TCR) to discriminate foreign from self-peptides presented by major histocompatibility complex (MHC) class II molecules is essential for an effective adaptive immune response. TCR recognition of self-peptides has been linked to autoimmune disease. Mutant self-peptides have been associated with tumors. Engagement of TCRs by a family of bacterial toxins know as superantigens has been responsible for toxic shock syndrome. Autoantibodies to V beta segments of T cell receptors have been isolated from patients with rheumatoid arthritis (RA) and systemic lupus erythematosus (SLE). The autoantibodies block TH1-mediated inflammatory autodestructive reactions and are believed to be a method by which the immune system compensates for disease. Most human T cells express the TCR alpha-beta and either CD4 or CD8 molecule (single positive, SP). A small number of T cells lack both CD4 and CD8 (double negative, DN). Increased percentages of alpha-beta DN T cells have been identified in some autoimmune and immunodeficiency disorders. Gamma-delta T cells are primarily found within the epithelium. They show less TCR diversity and recognize antigens differently than alpha-beta T cells. Subsets of gamma-delta T cells have shown antitumor and immunoregulatory activity.
Specifications
TCR V alpha 11 Chimeric | |
Recombinant Monoclonal | |
1 mg/mL | |
PBS with 0.02% ProClin 300 | |
Q05685 | |
Rabbit | |
Protein A | |
RUO | |
14276 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
ELISA, Flow Cytometry, Western Blot | |
34 | |
Unconjugated | |
FOLR2 | |
TCR V alpha11; TCRV alpha 11; TCRV alpha11; Va11; Valpha11 | |
Purified recombinant soluble TCR from the T-cell hybridoma 85.33, corresponding to V alpha 11. | |
200 μg | |
Primary | |
Human | |
Antibody | |
IgG κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction