Learn More
Invitrogen™ TCR V alpha 11 Monoclonal Antibody (RR8-1), APC, eBioscience™

Rat Monoclonal Antibody
$359.00
Specifications
| Antigen | TCR V alpha 11 |
|---|---|
| Clone | RR8-1 |
| Concentration | 0.2 mg/mL |
| Content And Storage | 4°C, store in dark, DO NOT FREEZE! |
| Applications | Flow Cytometry |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-245-695
![]()
|
Invitrogen™
17580082 |
100 μg |
Each of 1 for $359.00
|
|
|||||
Description
Description: This RR8-1 monoclonal antibody reacts with the mouse T cell receptor (TCR) V alpha 11 chain. Composed of an alpha and beta chain, TCR specificity is typically determined by Va, Ja, Vb, Db, and Jb gene rearrangement. The RR8-1 antibody recognizes the V beta 11 chain on T cells from mouse strains with the b (e.g., C57BL/6, B10) and d (e.g., DBA/1, DBA/2) haplotypes. V alpha 11+ T cells are absent in strains with the a (e.g., BALB/c, C3H) or c (e.g., SJL) haplotypes of the Tcra gene complex. Finally, V alpha 11 is expressed predominantly on CD4+ T cells. The RR8-1 antibody recognizes the V alpha 11.1 and 11.2 alleles, but not 11.3. This antibody has been reported to stimulate V alpha 11+ T cells. Applications Reported: This RR8-1 antibody has been reported for use in flow cytometric analysis. Applications Tested: This RR8-1 antibody has been tested by flow cytometric analysis of mouse lymph node. This can be used at less than or equal to 1 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Excitation: 633-647 nm; Emission: 660 nm; Laser: Red Laser. Filtration: 0.2 μm post-manufacturing filtered.
The ability of T cell receptors (TCR) to discriminate foreign from self-peptides presented by major histocompatibility complex (MHC) class II molecules is essential for an effective adaptive immune response. TCR recognition of self-peptides has been linked to autoimmune disease. Mutant self-peptides have been associated with tumors. Engagement of TCRs by a family of bacterial toxins know as superantigens has been responsible for toxic shock syndrome. Autoantibodies to V beta segments of T cell receptors have been isolated from patients with rheumatoid arthritis (RA) and systemic lupus erythematosus (SLE). The autoantibodies block TH1-mediated inflammatory autodestructive reactions and are believed to be a method by which the immune system compensates for disease. Most human T cells express the TCR alpha-beta and either CD4 or CD8 molecule (single positive, SP). A small number of T cells lack both CD4 and CD8 (double negative, DN). Increased percentages of alpha-beta DN T cells have been identified in some autoimmune and immunodeficiency disorders. Gamma-delta T cells are primarily found within the epithelium. They show less TCR diversity and recognize antigens differently than alpha-beta T cells. Subsets of gamma-delta T cells have shown antitumor and immunoregulatory activity.Specifications
| TCR V alpha 11 | |
| 0.2 mg/mL | |
| Flow Cytometry | |
| APC | |
| Rat | |
| RUO | |
| PBS with 0.09% sodium azide; pH 7.2 | |
| Primary | |
| Affinity chromatography |
| RR8-1 | |
| 4°C, store in dark, DO NOT FREEZE! | |
| Monoclonal | |
| Liquid | |
| IgG2b κ | |
| Mouse | |
| TCR V alpha11; TCRV alpha 11; TCRV alpha11; Va11; Valpha11 | |
| Antibody |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.