Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TCR V alpha 12.1 Antibody (6D6.6), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP222491
Description
TCR V alpha 12.1 Monoclonal specifically detects TCR V alpha 12.1 in Human, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Frozen.Specifications
TCR V alpha 12.1 | |
Monoclonal | |
0.15 mg/mL | |
Western Blot 1:1 - 1:100, Flow Cytometry 1 - 2 ug, Immunohistochemistry 1:10 - 1:500, Immunoprecipitation 1:10 - 1:500, Immunohistochemistry-Frozen 1:10 - 1:500, Functional | |
T cell receptor alpha locus, T-cell antigen receptor, alpha polypeptide, T-cell receptor, alpha cluster, TCR V alpha 12 TCRA, TCRD, TRA | |
Mouse | |
Protein G purified | |
RUO | |
6955 | |
Human, Rat | |
Ascites |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Frozen), Functional Assay | |
6D6.6 | |
Unconjugated | |
P01848 | |
TRA | |
Human TCR V alpha 12.1 | |
200 μg | |
Primary | |
This antibody reacts with Human. | |
Store at -20C. Avoid freeze-thaw cycles. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction