Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TCR V alpha 2 Monoclonal Antibody (B20.1), NovaFluor™ Yellow 610, eBioscience™, Invitrogen™
Rat Monoclonal Antibody
$194.00 - $375.00
Specifications
Antigen | TCR V alpha 2 |
---|---|
Clone | B20.1 |
Concentration | 0.1 mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
M033T03Y03A
![]() |
Invitrogen™
M033T03Y03A |
100 μg |
Each for $375.00
|
|
|||||
M033T02Y03A
![]() |
Invitrogen™
M033T02Y03A |
25 μg |
Each for $194.00
|
|
|||||
Description
TCR V alpha 2 Monoclonal antibody specifically detects TCR V alpha 2 in Mouse samples. It is validated for Flow CytometrySpecifications
TCR V alpha 2 | |
0.1 mg/mL | |
Monoclonal | |
Rat | |
Mouse | |
TCRVA; TCRVA2; V alpha 2; V alpha-2 TCR; Va 2; Va2; Valpha 2; Valpha2 | |
Primary | |
4°C, store in dark, DO NOT FREEZE! |
B20.1 | |
Flow Cytometry | |
NovaFluor Yellow 610 | |
RUO | |
PBS with 0.09% sodium azide; pH 7.2 | |
IgG2a λ | |
Affinity chromatography |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title