Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TCR V beta 10 Monoclonal Antibody (G101)
GREENER_CHOICE

Catalog No. PIMA180414
Change view
Click to view available options
Quantity:
250 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA180414 250 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA180414 Supplier Invitrogen™ Supplier No. MA180414
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

For FACS analysis, use 10 μL of the suggested working dilution to label 1x10^6 cells in 100 μL. Mouse anti Rat TCR V Beta 10 antibody, clone G101 recognizes the T-cell receptor V beta 10 gene product.

TRUSTED_SUSTAINABILITY

Specifications

Antigen TCR V beta 10
Applications Flow Cytometry
Classification Monoclonal
Clone G101
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.2
Gene Tcrb
Gene Accession No. 0
Gene Alias FLJ22602; MGC117436; MGC22624; MGC23964; MGC71411; RATTCB; RATTCBC1; T cell receptor beta locus; TCB; TCBC1; T-cell receptor beta chain; TCRA; TCRB; TCRD; TRA; TRB; TRB@
Gene Symbols Tcrb
Host Species Mouse
Immunogen T cell clone expressing the TCR V beta 10 gene product.
Purification Method Ion-exchange chromatography
Quantity 250 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 24820
Target Species Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.