Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TCR V beta 10 Antibody (G101) - BSA Free, Novus Biologicals™
SDP

Catalog No. p-7199352 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
125 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB001716 125 μg
NB017561 0.025 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB001716 Supplier Novus Biologicals Supplier No. NB10064281125UG
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

TCR V beta 10 Monoclonal specifically detects TCR V beta 10 in Rat samples. It is validated for Flow Cytometry.

Specifications

Antigen TCR V beta 10
Applications Flow Cytometry
Classification Monoclonal
Clone G101
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Flow Cytometry 1:50-1:100
Formulation PBS
Gene Alias T cell receptor beta locus, T-cell antigen receptor, beta polypeptide, T-cell receptor, beta cluster, T-cell receptor, beta cluster, TCRB, TRB
Gene Symbols TRB
Host Species Mouse
Immunogen T cell clone expressing the TCR V beta 10 gene product
Quantity 125 μg
Gene ID (Entrez) 6957
Test Specificity NB100-64281 is specific for the T-cell receptor V beta 10 gene product. Approximately 4.2% of unseparated LEW lymph node cells were detected by this clone. Reactivity is restricted to alpha/beta T cells, of which 6.7% labelled (1).
Target Species Rat
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.