Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TCR V beta 12 Monoclonal Antibody (S511)
GREENER_CHOICE

Catalog No. PITCR1654
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PITCR1654 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PITCR1654 Supplier Invitrogen™ Supplier No. TCR1654
Add to Cart
Edge
Add to Cart

Mouse Monoclonal Antibody

TCR1654 targets TCR V beta 12 in FACS, IHC(F), IP, and TCA applications and shows reactivity with Human samples. The TCR1654 immunogen is human TCR V beta 12.

The ability of T cell receptors (TCR) to discriminate foreign from self-peptides presented by major histocompatibility complex (MHC) class II molecules is essential for an effective adaptive immune response. TCR recognition of self-peptides has been linked to autoimmune disease. Mutant self-peptides have been associated with tumors. Engagement of TCRs by a family of bacterial toxins know as superantigens has been responsible for toxic shock syndrome. Autoantibodies to V beta segments of T cell receptors have been isolated from patients with rheumatoid arthritis (RA) and systemic lupus erythematosus (SLE). The autoantibodies block TH1-mediated inflammatory autodestructive reactions and are believed to be a method by which the immune system compensates for disease (ref5). T Cell and TCR Diversity Most human T cells express the TCR alpha-beta and either CD4 or CD8 molecule (single positive, SP). A small number of T cells lack both CD4 and CD8 (double negative, DN). Increased percentages of alpha-beta DN T cells have been identified in some autoimmune and immunodeficiency disorders. Gamma-delta T cells are primarily found within the epithelium. They show less TCR diversity and recognize antigens differently than alpha-beta T cells. Subsets of gamma-delta T cells have shown antitumor and immunoregulatory activity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TCR V beta 12
Applications Flow Cytometry, Immunohistochemistry (Frozen), Immunoprecipitation, T-Cell Activation
Classification Monoclonal
Clone S511
Concentration 0.15 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.5% BSA and 0.1% sodium azide
Gene TRB
Gene Accession No. P04435
Gene Alias T cell receptor beta chain; T cell receptor beta locus; TCRB; TCRbeta; Tib; TRB; TRB@
Gene Symbols TRB
Host Species Mouse
Immunogen Human TCR V beta 12.
Purification Method Protein A
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6957
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.