Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TCR V beta 5.3 Monoclonal Antibody (MEM-262)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA544006
This item is not returnable.
View return policy
Description
This antibody recognizes an extracellular epitope on beta chains of the TCR expressed by HPB-ALL cell line [carrying V(beta5.3)] and a small subset of peripheral blood T cells. This subset is larger than that recognized by other V(beta5.3)-specific antibodies.
The TCR V beta 5.3 antibody recognizes beta chains of the TCR expressed by HPB-ALL cell line carrying V beta 5.3 and a small subset of peripheral blood T cells. This subset is larger than that recognized by other V (beta5.3) specific antibodies.
Specifications
TCR V beta 5.3 | |
Monoclonal | |
1 mg/mL | |
PBS with 15mM sodium azide; pH 7.4 | |
P0DSE2 | |
TRB | |
Human thymoma cell line HPB-ALL. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2a |
Flow Cytometry, Immunoprecipitation, Western Blot | |
MEM-262 | |
Unconjugated | |
TRB | |
T cell receptor beta locus; TCRB; TRB; TRB@ | |
Mouse | |
Protein A | |
RUO | |
6957 | |
4°C, do not freeze | |
Liquid |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction