Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TCR V beta 5.3 Monoclonal Antibody (MEM-262)
GREENER_CHOICE

Catalog No. PIMA528598
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA528598 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA528598 Supplier Thermo Scientific Supplier No. MA528598
Add to Cart
Add to Cart

TCR V beta 5.3 Monoclonal Antibody (MEM-262)

TCR V beta 5.3 Monoclonal Antibody for Western Blot, Flow, IP, FN

TRUSTED_SUSTAINABILITY

Specifications

Antigen TCR V beta 5.3
Applications Flow Cytometry, Functional Assay, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone MEM-262
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene TRB
Gene Accession No. P0DSE2
Gene Alias T cell receptor beta locus; TCRB; TRB; TRB@
Gene Symbols TRB
Host Species Mouse
Immunogen Human thymoma cell line HPB-ALL.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6957
Target Species Human
Content And Storage 4°C, do not freeze
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.