Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TCR V beta 5 Antibody (MEM-262), DyLight 405, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB500520V
Description
TCR V beta 5 Monoclonal specifically detects TCR V beta 5 in Human samples. It is validated for Western Blot, Flow Cytometry.Specifications
| TCR V beta 5 | |
| Monoclonal | |
| DyLight 405 | |
| T cell receptor beta locus, T-cell antigen receptor, beta polypeptide, T-cell receptor, beta cluster, T-cell receptor, beta cluster, TCRB, TRB | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Primary | |
| The antibody MEM-262 recognizes beta chains of the TCR expressed by HPB-ALL cell line [carrying V(beta5.3)] and a small subset of peripheral blood T cells. This subset is larger than that recognized by other V(beta5.3)-specific antibodies. | |
| Store at 4C in the dark. |
| Western Blot, Flow Cytometry | |
| MEM-262 | |
| Western Blot, Flow Cytometry | |
| TRB | |
| Human thymoma cell line HPB-ALL. | |
| 0.1 mL | |
| Adaptive Immunity, Breast Cancer, Cancer, Cell Cycle and Replication, DNA Repair, Immunology | |
| 6957 | |
| Human | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction