Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TCR V beta 5 Antibody (MEM-262), PE/Cy7, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB500520PECY7
Description
TCR V beta 5 Monoclonal specifically detects TCR V beta 5 in Human samples. It is validated for Flow Cytometry.Specifications
| TCR V beta 5 | |
| Monoclonal | |
| PE-Cyanine7 | |
| PBS with 0.05% Sodium Azide | |
| TRB | |
| Human thymoma cell line HPB-ALL. | |
| 0.1 mL | |
| Primary | |
| The antibody MEM-262 recognizes beta chains of the TCR expressed by HPB-ALL cell line [carrying V(beta5.3)] and a small subset of peripheral blood T cells. This subset is larger than that recognized by other V(beta5.3)-specific antibodies. | |
| Store at 4°C in the dark. Do not freeze. |
| Flow Cytometry | |
| MEM-262 | |
| Flow Cytometry | |
| T cell receptor beta locus, T-cell antigen receptor, beta polypeptide, T-cell receptor, beta cluster, T-cell receptor, beta cluster, TCRB, TRB | |
| Mouse | |
| Protein A purified | |
| Adaptive Immunity, Breast Cancer, Cancer, Cell Cycle and Replication, DNA Repair, Immunology | |
| 6957.0 | |
| Human | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction