Learn More
Invitrogen™ TCR V beta 9 Monoclonal Antibody (MR10-2), eFluor™ 450, eBioscience™

Description
Description: This MR10-2 monoclonal antibody reacts with the mouse T-cell receptor (TCR) V beta 9 chain. Composed of an a and β chain, TCR specificity is typically determined by Va, Ja, Vb, Db, and Jb gene rearrangement. The MR10-2 antibody recognizes the V beta 9 chain on T cells from mouse strains with the b haplotype, including BALB/c, CBA/Ca, C3H/He, and C57BL. This gene is deleted in mice bearing the a (e.g., C57BR, C57L, SJL, and SWR) or c (e.g, RIII) haplotypes. T cells expressing V beta 9 are also deleted in mice expressing the minor lymphocyte stimulating (MIs)-1a antigen. The MR10-2 antibody has been reported to activate V beta 9+ T cells. Applications Reported: Anti-Mouse V beta 9 TCR eFluor® 450 has been reported for use in flow cytometric analysis. Applications Tested: Anti-Mouse V beta 9 TCR eFluor® 450 has been tested by flow cytometric analysis of mouse lymph node. This can be used at less than or equal to 0.5 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. eFluor® 450 is an alternative to Pacific Blue®. eFluor® 450 emits at 445 nm and is excited with the Violet laser (405 nm). Please make sure that your instrument is capable of detecting this fluorochome. Ex...
Specifications
Specifications
| Antigen | TCR V beta 9 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | MR10-2 |
| Concentration | 0.2 mg/mL |
| Conjugate | eFluor 450 |
| Formulation | PBS with 0.09% sodium azide; pH 7.2 |
| Gene Alias | TCR V beta9; TCRV beta 9; TCRV beta9; Vb9; Vbeta9 |
| Host Species | Mouse |
| Purification Method | Affinity chromatography |
| Show More |
Frequently Asked Questions (FAQs)
Our options will depend on the samples you are analyzing.
If cell viability is not critical, you can store your stained samples at 4 degrees C or on ice overnight in the dark and analyze the following day.
For samples stained with eFluor organic fluorochromes, we recommend that cells be suspended in 100 uL of Flow Cytometry Staining Buffer (Cat. No. 00-4222) and 100 uL of eBioscience IC Fixation Buffer (Cat. No. 00-8222); samples can be incubated for up to 3 days at 4 degrees C in the dark. Alternatively, the 1-step Fix/Lyse Solution (Cat. No. 00-5333) can be used. This is a great option when working with whole blood but also works for other cell types.
The eFluor Organic fluorochromes and eVolve QDots can be used with flow staining buffers containing PBS and protein.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.