Learn More
TCR V delta 1 Monoclonal Antibody (TS8.2), PE, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
$483.00
Specifications
Antigen | TCR V delta 1 |
---|---|
Clone | TS8.2 |
Concentration | 5 μL/Test |
Applications | Flow Cytometry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
12-567-942
![]() |
Invitrogen™
12567942 |
100 Tests |
Each of 1 for $483.00
|
|
|||||
Description
Description: This TS8.2 monoclonal antibody recognizes human TCR V delta 1, a common framework determinant on the delta-subunit of the T cell receptor. Gamma-delta T cells are primarily found within the epithelium. They show less TCR diversity and recognize antigens differently than alpha-beta T cells. Subsets of gamma-delta T cells have shown antitumor and immunoregulatory activity. TCR V delta 1 Monoclonal Antibody (TS8.2) has been validated for the following applications: Flow Cytometry. Applications Reported: This TS8.2 antibody has been reported for use in flow cytometric analysis. Applications Tested: This TS8.2 antibody has been pre-diluted and tested by flow cytometric analysis of normal human peripheral blood cells. This may be used at 5 μL (0.25 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Excitation: 488-561 nm; Emission: 578 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser
The T Cell Receptor gamma-delta (TCR gamma delta) is a heterodimeric membrane receptor that consists of the highly variable gamma and delta chains expressed as part of a complex with the invariant CD3 proteins. Gamma-delta T cells are less frequent than alpha-beta T cells but compose a substantial part of T cells found at epithelial borders including the gut mucosa resident intraepithelial lymphocytes (IEL). The antigenic stimuli required for gamma-delta T cell receptor activation are not well understood and are thought to be mainly MHC class-independent.Specifications
TCR V delta 1 | |
5 μL/Test | |
Monoclonal | |
Liquid | |
RUO | |
6964 | |
Full length protein of Human TCR delta v1. | |
Primary | |
4°C, store in dark, DO NOT FREEZE! | |
TRD |
TS8.2 | |
Flow Cytometry | |
PE | |
Mouse | |
Human | |
TRD | |
IgG1 κ | |
Affinity chromatography | |
Antibody |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.