Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TCR V delta 2 Antibody (B6), mFluor Violet 610 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP311706MFV610
Description
TCR V delta 2 Monoclonal antibody specifically detects TCR V delta 2 in Human samples. It is validated for Flow CytometrySpecifications
TCR V delta 2 | |
Monoclonal | |
mFluor Violet 610 SE | |
50mM Sodium Borate | |
Mouse | |
Protein A purified | |
RUO | |
28517 | |
Store at 4C in the dark. | |
IgG1 κ |
Flow Cytometry | |
B6 | |
Flow Cytometry | |
hDV102S1 | |
Human recombinant TCR Vdelta2 | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction