Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TCR VA7.2 Mouse, anti-Human, R718, Clone: OF-5A12, BD OptiBuild™

Catalog No. BDB751905 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB751905 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB751905 Supplier BD Biosciences Supplier No. 751905
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The OF-5A12 monoclonal antibody specifically recognizes the human variable alpha 7.2 region of the T cell receptor for antigen (TCR Vα7.2) that is joined with the Jα33 segment to form an invariant TCR which is characteristically expressed by mucosal-associated invariant T cells (MAIT cells). MAIT cell recognition of antigen is restricted by MHC-related molecule 1 (MR1), a nonpolymorphic Class Ib Major Histocompatibility Complex (MHC) molecule, that associates with β2-microglobulin. MAIT cells are found in human peripheral blood, mesenteric lymph nodes, liver, and mucosa associated with the lungs and intestines. MAIT cells play a role in detecting and fighting off microbial infections. The specificity of OF-5A12 for the TCR Vα7.2 was confirmed by its ability to block the binding of another human TCR Vα7.2-specific antibody, 3C10, to MAIT cells.

TRUSTED_SUSTAINABILITY

Specifications

Antigen TCR VA7.2
Applications Flow Cytometry
Classification Monoclonal
Clone OF-5A12
Conjugate R718
Host Species Mouse
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Refrigerate Upon Arrival
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.