Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TCRG/TCRD Hamster anti-Mouse, FITC, Clone: GL-3, Abnova™

Catalog No. 89934989 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-934-989 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-934-989 Supplier Abnova Corporation Supplier No. MAB12644
Add to Cart
Add to Cart

Hamster monoclonal antibody raised against mouse Tcrg/Tcrd.

Specifications

Antigen TCRG/TCRD
Applications Flow Cytometry, Immunohistochemistry (Frozen), Immunohistochemistry (PFA fixed)
Classification Monoclonal
Clone GL-3
Conjugate FITC
Description Hamster monoclonal antibody raised against mouse Tcrg/Tcrd.
Dilution Flow Cytometry (1 ug/106 cells) Immunohistochemistry (Frozen sections) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:50) The optimal working dilution should be determined by the end user.
Formulation In PBS (EIA grade BSA, 0.02% sodium azide).
Gene Alias TCRGV1S1/TCRGV2S1/TCRGV3S1/TCRGV5S3
Gene Symbols Tcrg
Host Species Hamster
Immunogen C57BL/6 intraepithelial lymphocytes.
Purification Method Protein G purification
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 110067/110066
Target Species Mouse
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.