Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ TCS 2510
GREENER_CHOICESDP

Catalog No. 40691
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
40-691 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 40-691 Supplier Tocris Bioscience™ Supplier No. 4069/1

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Selective EP4 agonist

Highly selective EP4 agonist (EC50 = 2.5 nM; Ki = 1.2 nM). Displays no significant binding at other prostaglandin receptors at concentrations up to 14 μM.

Chemical Identifiers

CAS 346673-06-1
Molecular Formula C21H29N5O2

Specifications

Quantity 1 mg
Formula Weight 383.49
Purity >98%
Molecular Weight (g/mol) 383.496
Chemical Name or Material TCS 2510
Recommended Storage Store at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.