Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ TCS ERK 11e
GREENER_CHOICESDP

Catalog No. 446510 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
44-651-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 44-651-0 Supplier Tocris Bioscience™ Supplier No. 4465/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Potent and selective ERK2 inhibitor

Potent and selective extracellular signal-related kinase 2 (ERK2) inhibitor (Ki values are <2, 395, 540 and 852 nM for ERK2, GSK-3, Aurora Kinase A and Cdk2 respectively). Potently blocks proliferation of HT29 cells (IC50 = 48 nM). Orally bioavailable.

Chemical Identifiers

CAS 896720-20-0
Molecular Formula C24H20Cl2FN5O2

Specifications

Quantity 10 mg
Formula Weight 500.35
Purity >98%
Molecular Weight (g/mol) 500.355
Chemical Name or Material TCS ERK 11e
Recommended Storage Store at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.