Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ TCS JNK 5a
GREENER_CHOICESDP

Catalog No. 282750 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
28-275-0 50 mg
28-271-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 28-275-0 Supplier R&D Systems™ Supplier No. 2827/50

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Selective inhibitor of JNK2 and JNK3

Selective inhibitor of JNK2 and JNK3 (pIC50 values are 6.7, 6.5, <5.0 and <4.8 for JNK3, JNK2, JNK1 and p38α respectively). Displays no significant activity at a range of other protein kinases including EGFR, ErbB2, cdk2, PLK-1 and Src (pIC50 < 5.0).

Chemical Identifiers

CAS 312917-14-9
Molecular Formula C20H16N2OS

Specifications

Quantity 50 mg
Product Line Tocris
Formula Weight 332.42
Purity >99%
Molecular Weight (g/mol) 332.421
Chemical Name or Material TCS JNK 5a
Recommended Storage Store at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.