Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ TCS OX2 29
GREENER_CHOICESDP

Catalog No. 337150 Shop All Novus Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
33-715-0 50 mg
33-711-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 33-715-0 Supplier R&D Systems™ Supplier No. 3371/50

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Potent and selective OX2 antagonist

Potent and selective OX2 receptor antagonist (IC50 = 40 nM). Displays >250-fold selectivity for OX2 over OX1 and a range of receptors, ion channels and transporters. Inhibits orexin A induced IP3 accumulation and ERK1/2 phosphorylation in CHO cells transfected with the OX2 receptor.

Chemical Identifiers

CAS 1610882-30-8
Molecular Formula C23H32ClN3O3

Specifications

Quantity 50 mg
Product Line Tocris
Formula Weight 433.97
Purity >98%
Molecular Weight (g/mol) 433.977
Chemical Name or Material TCS OX2 29
Recommended Storage Desiccate at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.