Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TCTA Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5120347
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5120347 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5120347 Supplier Invitrogen™ Supplier No. PA5120347
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive test controls include: 293T, MCF7, SGC-7901. The target is usually found in the following locations: Membrane, Multi-pass membrane protein. Immunogen sequence: MAESWSGQAL QALPATVLGA LGSEFLREWE AQDMRVTLFK LLLLWLVLSL LGIQLAWGFY GNTVTGLYHR PGLGGQNGST PDGSTHFPSW EMAANEPLKT HRE.

May be required for cellular fusion during osteoclastogenesis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TCTA
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 2.72 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene TCTA
Gene Accession No. P57738, Q5XIF1, Q8VEA7
Gene Alias 9130410M22Rik; AW553637; C85065; T cell leukemia translocation altered gene; T-cell leukemia translocation altered; T-cell leukemia translocation-altered gene protein; T-cell leukemia translocation-altered gene protein homolog; T-cell leukemia translocation-associated gene protein; Tcta; Tctal
Gene Symbols TCTA
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-103 of human TCTA (NP_071503.1).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 102791, 306587, 6988
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.