Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TCTA Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA552424
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA552424 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA552424 Supplier Invitrogen™ Supplier No. PA552424
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: AESWSGQALQ ALPATVLGAL GSEFLREWEA QDMRVTLFKL Highest antigen sequence identity to the following orthologs: Mouse - 81%, Rat - 84%.

May be required for cellular fusion during osteoclastogenesis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TCTA
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TCTA
Gene Accession No. P57738
Gene Alias 9130410M22Rik; AW553637; C85065; T cell leukemia translocation altered gene; T-cell leukemia translocation altered; T-cell leukemia translocation-altered gene protein; T-cell leukemia translocation-altered gene protein homolog; T-cell leukemia translocation-associated gene protein; Tcta; Tctal
Gene Symbols TCTA
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human TCTA. Recombinant protein control fragment (Product #RP-88687).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6988
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.