Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TCTE1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5112963
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5112963 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5112963 Supplier Invitrogen™ Supplier No. PA5112963
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TCTE1 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), ELISA

TCTE1 is a protein coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TCTE1
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene TCTE1
Gene Accession No. A6H639, Q5JU00
Gene Alias D17sil1; D6S46; Drc5; Dynein regulatory complex subunit 5; FAP155; t-complex-associated testis expressed 1; T-complex-associated testis-expressed protein 1; t-complex-associated-testis-expressed 1; Tcte1; tcte-1
Gene Symbols TCTE1
Host Species Rabbit
Immunogen Recombinant Human Dynein regulatory complex subunit 5 protein (285-501AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 202500, 21645
Target Species Human, Mouse
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.