Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TCTEX1D2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA546636
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA546636 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA546636 Supplier Invitrogen™ Supplier No. PA546636
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: ELANAEYSPE EMPQLTKHLS ENIKDKLKEM GFDRYKMVVQ VVIGEQRGEG Sequence homology: Cow: 100%; Dog: 86%; Guinea Pig: 100%; Horse: 93%; Human: 100%; Mouse: 86%; Rabbit: 86%; Rat: 86%; Yeast: 82%.

Dyneins are multi-subunit, high molecular weight ATPases that interact with microtubules to generate force by converting the chemical energy of ATP into the mechanical energy of movement. Cytoplasmic or axonemal Dynein is an approximately 12 subunit complex of two heavy chains, two intermediate chains to anchor Dynein to its cargo, four smaller intermediate chains and several light chains. It performs functions necessary for cell survival such as organelle transport and centrosome assembly. TCTEX1 is a cytoplasmic dynein light chain found in a complex with Na+ CP type X (SCN10A). TCTEX1D2, also called TCTEX1 domain containing 2, belongs to the dynein light chain TCTEX-type family. Its function is still under investigation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TCTEX1D2
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene TCTEX1D2
Gene Accession No. Q8WW35
Gene Alias 0610012D17Rik; 2410011K15Rik; dynein light chain Tctex-type 2B; DYNLT2B; RGD1563597; Tctex1 domain containing 2; tctex1 domain-containing protein 2; TCTEX1D2; Unknown (protein for IMAGE:8040601)
Gene Symbols TCTEX1D2
Host Species Rabbit
Immunogen Synthetic peptide directed towards the C-terminal of human TCTEX1D2 (aa 55-104).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 255758
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.